missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx2.4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567368
This item is not returnable.
View return policy
Description
Immunogen sequence: RYSSISRFMGP SAGVNVAGMG SLTGIADAAK SLGPL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
NKX2-4(NK2 Homeobox4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includesequence-specific DNA binding and may have transcription factor function.
Specifications
| Nkx2.4 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-4 | |
| 1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 644524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2Z4 | |
| NKX2-4 | |
| Recombinant Human Nkx2.4. Recombinant protein control fragment (Product #RP-108025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction