missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx2.4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101641
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NKX24.
NKX2-4(NK2 Homeobox4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includesequence-specific DNA binding and may have transcription factor function.
Specifications
| Nkx2.4 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-4 | |
| 1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 644524 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H2Z4 | |
| NKX2-4 | |
| A synthesized peptide derived from human NKX2-4(Accession Q9H2Z4), corresponding to amino acid residues T92-D142. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction