missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Antibody (NX2/1524), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54323PE
This item is not returnable.
View return policy
Beskrivning
NKX2.2 Monoclonal antibody specifically detects NKX2.2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifikationer
| NKX2.2 | |
| Monoclonal | |
| PE | |
| Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
| Recombinant fragment (around aa1-119) of human NKX2.2 protein (exact sequence is proprietary) (Uniprot: O95096) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| NX2/1524 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4821 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering