missing translation for 'onlineSavingsMsg'
Learn More

NKX1-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-99300-100ul

 View more versions of this product

Product Code. 18694875

  • £255.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NKX1-2 Polyclonal antibody specifically detects NKX1-2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NKX1-2
Polyclonal
Western Blot 1:500-1:2000, Immunoprecipitation 5-10 uL/mg of lysate
bB238F13.2, C10orf121, NKX-1.1, NKX1-2 NK1 homeobox 2
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse NKX1-2.
100 μg
Primary
Mouse
Purified
Western Blot, Immunoprecipitation
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
390010
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.