missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKTR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115655
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NKTR.
This gene encodes a membrane-anchored protein with a hydrophobic amino terminal domain and a cyclophilin-like PPIase domain. It is present on the surface of natural killer cells and facilitates their binding to targets. Its expression is regulated by IL2 activation of the cells.
Specifications
| NKTR | |
| Polyclonal | |
| Unconjugated | |
| Nktr | |
| 5330401F18Rik; cyclophilin-related protein; D9Wsu172e; natural killer cell triggering receptor; natural killer triggering receptor; natural killer tumor recognition protein; natural killer tumor recognition sequence; natural killer tumor recognition sequence-like; natural killer-tumor recognition sequence; natural-killer cells cyclophilin-related protein; NKTR; NK-TR protein; NK-tumor recognition protein; p104; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100364165, 18087, 4820 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P30414, P30415 | |
| Nktr | |
| A synthesized peptide derived from human NKTR(Accession P30414), corresponding to amino acid residues E800-E850. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction