missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKTR Polyclonal Antibody

NKTR Polyclonal Antibody
Brand: Invitrogen PA565671
This item is not returnable.
View return policy
Description
NKTR Polyclonal Antibody for ICC/IF
Specifications
| NKTR | |
| Polyclonal | |
| Unconjugated | |
| Nktr | |
| 5330401F18Rik; cyclophilin-related protein; D9Wsu172e; natural killer cell triggering receptor; natural killer triggering receptor; natural killer tumor recognition protein; natural killer tumor recognition sequence; natural killer tumor recognition sequence-like; natural killer-tumor recognition sequence; natural-killer cells cyclophilin-related protein; NKTR; NK-TR protein; NK-tumor recognition protein; p104; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P30414 | |
| Nktr | |
| Recombinant Human NKTR. Recombinant protein control fragment (Product #RP-106346). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction