missing translation for 'onlineSavingsMsg'
Learn More

NKp80/KLRF1 Antibody (5D12), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18373303
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373303 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373303 Supplier Bio-Techne Supplier No. NBP314624AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKp80/KLRF1 Monoclonal antibody specifically detects NKp80/KLRF1 in Human, Monkey samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp80/KLRF1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 5D12
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
Host Species Mouse
Immunogen Recombinant human NKp80/KLRF1 extracellular domain
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51348
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.