missing translation for 'onlineSavingsMsg'
Learn More

NKp46/NCR1 Antibody (195314) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30168311 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30168311 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30168311 Supplier Novus Biologicals Supplier No. FAB1850JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Agonist Activity,Immunocytochemistry,CyTOF-reported,Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp46/NCR1
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Agonist Activity, Immunocytochemistry, CyTOF, Dual RNAScope ISH-IHC
Classification Monoclonal
Clone 195314
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp46/NCR1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9437
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.