missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp44/NCR2 Antibody (1G6), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42683PECY7
This item is not returnable.
View return policy
Description
NKp44/NCR2 Monoclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKp44/NCR2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| NCR2 | |
| Recombinant human NKp44 (19-130aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 1G6 | |
| Flow Cytometry | |
| CD336, CD336 antigen, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1 | |
| Mouse | |
| Protein A purified | |
| Innate Immunity | |
| 9436.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction