missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKp30 Recombinant Rabbit Monoclonal Antibody (4P2B3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558107
This item is not returnable.
View return policy
Description
Immunogen sequence: LWVSQPPEIR TLEGSSAFLP CSFNASQGRL AIGSVTWFRD EVVPGKEVRN GTPEFRGRLA PLASSRFLHD HQAELHIRDV RGHDASIYVC RVEVLGLGVG TGNGTRLVVE KEHPQLG.
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
Specifications
| NKp30 | |
| Recombinant Monoclonal | |
| 1.32 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O14931 | |
| Ncr3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-135 of human NCR3 (NP_667341.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 4P2B3 | |
| Unconjugated | |
| Ncr3 | |
| 1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED | |
| Rabbit | |
| Protein A | |
| RUO | |
| 259197 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction