missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30/NCR3 Antibody (210845), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1849J
This item is not returnable.
View return policy
Description
NKp30/NCR3 Monoclonal antibody specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF
Specifications
| NKp30/NCR3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Agonist Activity, CyTOF | |
| 210845 | |
| Flow Cytometry, Agonist Activity, CyTOF-ready | |
| Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS | |
| Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera | |
| 0.1 mL | |
| Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells | |
| 259197 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction