missing translation for 'onlineSavingsMsg'
Learn More

NKp30/NCR3 Antibody (012), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18394361 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394361 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394361 Supplier Novus Biologicals Supplier No. NBP289525AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp30/NCR3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 12
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human NKp30/NCR3 (NP_667341.1; Met1-Gly135).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 259197
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.