missing translation for 'onlineSavingsMsg'
Learn More

NKP-1339, MedChemExpress

Product Code. 16473087
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473087 50 mg 50mg
16467038 10 mg 10mg
16427096 100 mg 100mg
16419587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473087 Supplier Medchem Express Supplier No. HY16350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NKP-1339 (IT-139; KP-1339) is the first-in-class ruthenium-based anticancer agent in development against solid cancer with limited side effects. NKP-1339 induces G2/M cell cycle arrest, blockage of DNA synthesis, and induction of apoptosis via the mitochondrial pathway. NKP-1339 has a high tumor targeting potential, strongly binds to serum proteins such as albumin and transferrin and activates in the reductive tumor milieu.

NKP-1339 (IT-139

Specifications

Chemical Name or Material NKP-1339
Physical Form Solid
CAS 197723-00-5
Color Dark Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H12Cl4N4NaRu
For Use With (Application) Cancer-Kinase/protease
Synonym IT-139 KP-1339
Solubility Information DMSO : ≥ 59 mg/mL (117.50 mM)
SMILES [Cl-][Ru+3]([N]1=CC2=CC=CC=C2N1)([Cl-])([Cl-])([N]3=CC4=CC=CC=C4N3)[Cl-].[Na+]
Molecular Weight (g/mol) 502.14
Formula Weight 502.14
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.