missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIRAS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19545
This item is not returnable.
View return policy
Description
NKIRAS2 Polyclonal specifically detects NKIRAS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NKIRAS2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| DKFZP434N1526, I-kappa-B-interacting Ras-like protein 2, Kappa B-Ras protein 2, KappaB-Ras2, KBRAS2DKFZp434N1526, MGC74742, NF-kappa-B inhibitor-interacting Ras-like protein 2, NFKB inhibitor interacting Ras-like 2, NFKB inhibitor interacting Ras-like protein 2 | |
| Rabbit | |
| 22 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 28511 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.76mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9NYR9 | |
| NKIRAS2 | |
| Recombinant protein encompassing a sequence within the center region of human NKIRAS2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (100%), Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction