missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIRAS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06173-100ul
This item is not returnable.
View return policy
Description
NKIRAS1 Polyclonal antibody specifically detects NKIRAS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NKIRAS1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| I-kappa-B-interacting Ras-like protein 1, kappa B-ras 1, Kappa B-Ras protein 1, KappaB-Ras1, KBRAS1kappaB-Ras1, NF-kappa-B inhibitor-interacting Ras-like protein 1, NFKB inhibitor interacting Ras-like 1, NFKB inhibitor interacting Ras-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human NKIRAS1 fragment. | |
| 100 μg | |
| Signal Transduction | |
| 28512 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction