missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKIAMRE Polyclonal Antibody

Description
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (93%), Rat (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | NKIAMRE |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.03 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CDKL3 |
| Gene Accession No. | Q8IVW4 |
| Gene Alias | AI852479; B230379H01Rik; CDKL3; cyclin dependent kinase like 3; cyclin-dependent kinase-like 3; NKIAMRE; Nkiatre; Nkiatreb; serine/threonine kinase NKIATRE beta; serine/threonine protein kinase NKIATRE; Serine/threonine-protein kinase NKIAMRE; serine-threonine protein kinase NKIAMRE |
| Gene Symbols | CDKL3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?