missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIAMRE Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-122AF700
This item is not returnable.
View return policy
Description
NKIAMRE Polyclonal antibody specifically detects NKIAMRE in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| NKIAMRE | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 700 | |
| cyclin-dependent kinase-like 3, EC 2.7.11, EC 2.7.11.22, NKIAMRESerine/threonine-protein kinase NKIAMRE, serine-threonine protein kinase NKIAMRE | |
| Synthetic peptide corresponding to residues 14-29 [SYGTVMKCKHKNTGQI] of the 50 kDa human NKIAMRE protein. | |
| 0.1 mL | |
| Cancer, Protein Kinase, Tumor Suppressors | |
| 51265 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction