missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144131
This item is not returnable.
View return policy
Description
NKG7 Polyclonal Antibody for ICC/IF, ELISA
NKG7 is a protein coding gene.
Specifications
| NKG7 | |
| Polyclonal | |
| Unconjugated | |
| NKG7 | |
| G-CSF-induced gene 1 protein; GIG1; GIG-1 protein; GMP-17; granule membrane protein 17; granule membrane protein of 17 kDa; natural killer cell granule protein 7; natural killer cell group 7 sequence; natural killer cell protein 7; NKG7; p15-TIA-1; Protein NKG7 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 4818 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q16617 | |
| NKG7 | |
| Recombinant Human Protein NKG7 protein (30-60AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction