Learn More
NKG2E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA580761
Additional Details : Weight : 0.01000kg
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KLRC3/NKG2E.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity.Specifications
NKG2E | |
Polyclonal | |
Unconjugated | |
KLRC3 | |
killer cell lectin like receptor C3; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc2; KLRC3; natural killer cell group 2E cell receptor; NK cell receptor E; NKG2E; NKG2-E; NKG2-E type II integral membrane protein; NKG2-E-activating NK receptor | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
5 mg/mL | |
PBS with no preservative | |
Q07444 | |
KLRC3 | |
A synthetic peptide corresponding to the N-terminus of the Human KLRC3/NKG2E | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |