missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKG2E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564033
This item is not returnable.
View return policy
Description
Immunogen sequence: NCAMLHVRGL ISDQCGSSRI IRRGFIMLTR LVLN Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 35%.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity.
Specifications
| NKG2E | |
| Polyclonal | |
| Unconjugated | |
| KLRC3 | |
| killer cell lectin like receptor C3; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc2; KLRC3; natural killer cell group 2E cell receptor; NK cell receptor E; NKG2E; NKG2-E; NKG2-E type II integral membrane protein; NKG2-E-activating NK receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q07444 | |
| KLRC3 | |
| Recombinant protein corresponding to Human NKG2E. Recombinant protein control fragment (Product #RP-104534). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction