missing translation for 'onlineSavingsMsg'
Learn More

NKG2E/KLRC3 Antibody (3D5), Novus Biologicals™

Product Code. 18349378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349378 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349378 Supplier Novus Biologicals Supplier No. H00003823M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKG2E/KLRC3 Monoclonal antibody specifically detects NKG2E/KLRC3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG2E/KLRC3
Applications Western Blot, Flow Cytometry, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3D5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002252
Gene Alias killer cell lectin-like receptor subfamily C, member 3, NK cell receptor E, NKG2-E type II integral membrane protein, NKG2-E-activating NK receptor, NKG2ENKG2-E
Host Species Mouse
Immunogen KLRC3 (NP_002252, 132 a.a. ~ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GKERRTWEESLQACASKNSSSLLSIDNEEEMKFLASILPSSWIGVFRNSSHHPWVTINGLAFKHEIKDSDHAERNCAMLHVRGLISDQCGSSRIIRRGFIMLTRLVLNS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3823
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.