Learn More
NKG2D Mouse anti-Human, APC, Clone: 1D11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558071
Description
Antibody 1D11 reacts with NKG2D (42kDa), an activating receptor for MICA encoded within MHC. Different from natural cytotoxicity receptor (NCR), NKG2D expression is not confined to NK cells, it is also present on virtually all TCR γ/δ+ and CD8+TCR α/β+ T cells. NKG2D functions as a triggering receptor involved in natural cytotoxicity mediated by normal NK cells against a variety of tumors or normal target cells. More importantly, NKG2D can complement the role of NCR in tumor cell lysis. Remarkably, the combined maskings of NCR and NKG2D led to a complete inhibition of NK-mediated lysis of all tumor or normal cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 1D11
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
| NKG2D | |
| Monoclonal | |
| APC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1D11 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| NKL cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.