missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NKG2A/CD159a/KLRC1 Antibody (monalizumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28102-1mg
This item is not returnable.
View return policy
Description
NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| NKG2A/CD159a/KLRC1 | |
| Monoclonal | |
| Unconjugated | |
| P26715 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| monalizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| NKG2A/ CD94 | |
| 1 mg | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3821 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction