missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC2/SLC12A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£163.00
Specifications
| Antigen | NKCC2/SLC12A1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
NKCC2/SLC12A1 Polyclonal specifically detects NKCC2/SLC12A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| NKCC2/SLC12A1 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Rat | |
| Q5PQV1 | |
| 6557 | |
| Produced against a synthetic peptide mapping to a segment (amino acids 33-55) of the N-terminal tail of rat NKCC2 (antibody desgination L224). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| BSC1, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2, Kidney-specific Na-K-Cl symporter, MGC48843, NKCC2A variant A, NKCC2Na-K-2Cl cotransporter, solute carrier family 12 (sodium/potassium/chloride transporters), member 1, solute carrier family 12 member 1 | |
| SLC12A1 | |
| IgG | |
| Affinity Purified | |
| Detects approx 160kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title