missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC1 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577548
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
NKCC1 mediates sodium and chloride transport and reabsorption. NKCC1 is a membrane protein and is important in maintaining proper ionic balance and cell volume. NKCC1 is an electrically silent transporter system that mediates sodium and chloride reabsorption. Further, NKCC1 plays a vital role in the regulation of ionic balance and cell volume and is phosphorylated in response to DNA damage. Three transcript variants encoding two different isoforms have been found for NKCC1. Diseases associated with SLC12A2 include Basosquamous Carcinoma and Metatypical Basal Cell Carcinoma.
Specifications
| NKCC1 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| SLC12A2 | |
| NKCC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20496, 6558, 83629 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.6 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P55011, P55012 | |
| SLC12A2 | |
| Peptide (C)RRQAMKEMSIDQAK, corresponding to amino acid residues 828-841 of human NKCC1 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction