missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKAPL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£390.00
Specifications
| Antigen | NKAPL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NKAPL Polyclonal specifically detects NKAPL in Human samples. It is validated for Western Blot.Specifications
| NKAPL | |
| Polyclonal | |
| Rabbit | |
| MGC126734, NFKB activating protein-like | |
| NKAPL | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 222698 | |
| Synthetic peptides corresponding to NKAPL (NFKB activating protein-like) The peptide sequence was selected from the middle region of NKAPL)(50ug). Peptide sequence KTSRSRNKKKRKNKSSKRKHRKYSDSDSNSESDTNSDSDDDKKRVKAKKK. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title