missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551484
This item is not returnable.
View return policy
Description
Immunogen sequence: RRRSSSKSPK PSKSARSPRG RRSRSHSCSR SGDRNGLTHQ LGGLSQGSRN QSYRSRSRSR SRERPSAPRG IPFASASSSV YYGSYSRPYG SDKPWPSLLD KEREESLRQK RLS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
NKAP is a transcriptional repressor that associates with the NOTCH (see MIM 190198) corepressor complex and is required for T-cell development.
Specifications
| NKAP | |
| Polyclonal | |
| Unconjugated | |
| NKAP | |
| 2610020O08Rik; AA987160; AI849147; AL024236; c6orf194l; FLJ22626; FLJ22626-like; NF-kappaB activating protein; NF-kappa-B-activating protein; NF-kappa-B-activating protein; LOW QUALITY PROTEIN: NF-kappa-B-activating protein; NFKB activating protein; NKAP; nkapl; nuclear NF-kappaB activating protein; wu:fj36d09; wu:fj49c09; zgc:136458 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79576 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5F7 | |
| NKAP | |
| Recombinant protein corresponding to Human NKAP. Recombinant protein control fragment (Product #RP-91920). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction