missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115654
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NKAP.
NKAP is a transcriptional repressor that associates with the NOTCH (see MIM 190198) corepressor complex and is required for T-cell development.
Specifications
| NKAP | |
| Polyclonal | |
| Unconjugated | |
| NKAP | |
| 2610020O08Rik; AA987160; AI849147; AL024236; c6orf194l; FLJ22626; FLJ22626-like; NF-kappaB activating protein; NF-kappa-B-activating protein; NF-kappa-B-activating protein; LOW QUALITY PROTEIN: NF-kappa-B-activating protein; NFKB activating protein; NKAP; nkapl; nuclear NF-kappaB activating protein; wu:fj36d09; wu:fj49c09; zgc:136458 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 298342, 67050, 79576 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q4V7C9, Q8N5F7, Q9D0F4 | |
| NKAP | |
| A synthesized peptide derived from human NKAP(Accession Q8N5F7), corresponding to amino acid residues G6-L56. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction