missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKAIN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556280
This item is not returnable.
View return policy
Description
Immunogen sequence: LKDSELLTFS LSRHRSWWRE RWPGCLHEEV PAVGLGAPHG QALVSGAGCA LEPSYVEALH SCLQILIALL GFVCGCQVVS VFTEEEDSFD FIGGFDPFPL YHVNEKPSSL LSKQVY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.
NKAIN4 is a member of a family of mammalian proteins (see NKAIN1; MIM 612871) with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1; MIM 182330) (Gorokhova et al., 2007 [PubMed 17606467]).[supplied by OMIM, Jun 2009].
Specifications
| NKAIN4 | |
| Polyclonal | |
| Unconjugated | |
| NKAIN4 | |
| bA261N11.2; C20orf58; FAM77A; Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 4; Na+/K+ transporting ATPase interacting 4; NKAIN4; Protein FAM77A; Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVV8 | |
| NKAIN4 | |
| Recombinant protein corresponding to Human NKAIN4. Recombinant protein control fragment (Product #RP-90926). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction