missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK3R/TACR3/Neurokinin B Receptor Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06055MFV450
This item is not returnable.
View return policy
Description
NK3R/TACR3/Neurokinin B Receptor Polyclonal antibody specifically detects NK3R/TACR3/Neurokinin B Receptor in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NK3R/TACR3/Neurokinin B Receptor | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060 | |
| Produced in rabbits immunized with E. coli-derived Human NK3R/TACR3/Neurokinin B Receptor fragment. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6870 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction