missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NK2R Polyclonal antibody specifically detects NK2R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | NK2R |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | Neurokinin A receptor, NK2RNK-2 receptor, NKNARseven transmembrane helix receptor, SKRneurokinin 2 receptor, substance-K receptor, TAC2Rsubstance K receptor, tachykinin receptor 2NK-2R |
| Host Species | Rabbit |
| Immunogen | Synthetic 20 amino acid peptide from C-Terminus of human NK2R. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?