missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Rabbit anti-Human, Mouse, Rat, Clone: 9T1W2, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | NK1R |
|---|---|
| Clone | 9T1W2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326502
|
Novus Biologicals
NBP3-16461-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18364067
|
Novus Biologicals
NBP3-16461-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK1R Monoclonal antibody specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NK1R | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6869 | |
| IgG | |
| Affinity purified |
| 9T1W2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 300-407 of human NK1R (NK1R) (P25103). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title