missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Antibody, PE/Cy7, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-119PECY7
This item is not returnable.
View return policy
Description
NK1R Polyclonal specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| NK1R | |
| Polyclonal | |
| Flow Cytometry | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| Rabbit | |
| Affinity Purified | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6869.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TACR1 | |
| A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProt No. P25103] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction