missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
- 1.NB300-101: NK1R Antibody
- 2.NB300-119: NK1R Antibody
NK1R specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | NK1R |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor |
| Gene Symbols | TACR1 |
| Host Species | Rabbit |
| Immunogen | See individual datasheets. |
| Quantity | 2 Vials |
| Gene ID (Entrez) | 6869 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?