Learn More
NK1.1 (CD161) Mouse, anti-Mouse, FITC, Clone: PK136, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60103FI
Encompass_Preferred
 View more versions of this product
Product Code. 17199011
  • £504.90 / 500µg
Description
  • The PK136 antibody reacts with murine NK1.1 (CD161), an ∼80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice
  • NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively
  • CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, or 129)
  • NK1.1 has functional roles in modulating several processes, including the activation and proliferation of NK cells, induction of interferon-γ production, and release of cytotoxic granules
  • Its expression on the cell surface is specifically upregulated by IL-12
  • For detection of NK cells in NK1.1- strains, Anti-Mouse CD49b Antibody, Clone DX5 (Catalog #60020) is recommended
  • DX5 recognizes CD49b (integrin α2), another commonly used NK cell marker
Specifications
Specifications
NK1.1 (CD161)
Monoclonal
FITC
500 μg
Mouse
NK Cells
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
PK136
Mouse
Primary
IgG2a κ