missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NK-4R Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537272
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 46 kDa. Purity is >95% by SDS-PAGE.
NK4R is a putative Tachykinin Receptor with sequence similarities to the neuromedin K receptor. It was initially thought to be an opioid receptor and is antagonized by the opioid peptide dynorphin. However, NK4R is activated by the tachykinin neurokinin B.
Specifications
| NK-4R | |
| Polyclonal | |
| Unconjugated | |
| Nk4 receptor; NK4R; Tac3rl; Tacr4 | |
| A synthetic peptide corresponding to residues in human NK-4R. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction