Learn More
NK-1.1 Mouse anti-Mouse, PerCP-Cy5.5, Clone: PK136, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551114
Description
In the mouse, at least three members of the Klrb (Killer cell lectin-like receptor, subfamily b; formerly NKR-P1) gene family have been identified (Klrb1a/NKR-P1A, Klrb1b/NKR-P1B, and Klrb1c/NKR-P1C); but in the human gene family, a single homologue has b
Host Species: Mouse
Clone: PK136
Isotype: IgG2a κ
Species Reactivity: Mouse
Flow Cytometry
Specifications
| NK-1.1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| PK136 | |
| PerCP-Cyanine5.5 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse NK-1+ Spleen and Bone Marrow Cells | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.