missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK-1.1 Mouse anti-Mouse, BB700, Clone: PK136, BD Horizon™
Mouse Monoclonal Antibody
£118.64 - £374.56
Specifications
| Antigen | NK-1.1 |
|---|---|
| Clone | PK136 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15835109
|
BD Biosciences
566503 |
25 μg |
£125.00 £118.64 / Each Save £6.36 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15825109
|
BD Biosciences
566502 |
0.1 mg |
£391.00 £374.56 / Each Save £16.44 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK-1.1 Monoclonal antibody specifically detects NK-1.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| NK-1.1 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
| PK136 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title