missing translation for 'onlineSavingsMsg'
Learn More

NK-1.1 Mouse anti-Mouse, APC, Clone: PK136, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550627

 View more versions of this product

Product Code. 15876348

  • ONLINE EXCLUSIVE PRICE
    £283.00
    £271.18 / Each
    Save £11.82
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

In the mouse, at least three members of the Klrb (Killer cell lectin-like receptor, subfamily b; formerly NKR-P1) gene family have been identified (Klrb1a/NKR-P1A, Klrb1b/NKR-P1B, and Klrb1c/NKR-P1C); but in the human gene family, a single homologue has b

Host Species: Mouse
Clone: PK136
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse NK-1+ Spleen and Bone Marrow

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NK-1.1
Monoclonal
0.2mg/mL
Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
PK136
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse NK-1+ Spleen and Bone Marrow
0.1 mg
Immunology
Mouse
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.