missing translation for 'onlineSavingsMsg'
Learn More

Nizatidine Sulfoxide, TRC

Product Code. 30476730
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476730 100 mg -
30471287 25 mg 25mg
30473773 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476730 Supplier Toronto Research Chemicals Supplier No. TRCN598510100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102273-13-2
Molecular Formula C12 H21 N5 O3 S2
Molecular Weight (g/mol) 347.46
Synonym (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Nizatidine Imp. C (EP)
IUPAC Name 1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
SMILES CNC(=C[N+](=O)[O-])NCCS(=O)Cc1csc(CN(C)C)n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H21N5O3S2/c1-13-11(6-17(18)19)14-4-5-22(20)9-10-8-21-12(15-10)7-16(2)3/h6,8,13-14H,4-5,7,9H2,1-3H3
Formula Weight 347.1086
Chemical Name or Material (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.