missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitroxoline, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Nitroxoline
Specifications
Specifications
| Chemical Name or Material | Nitroxoline |
| CAS | 4008-48-4 |
| Analyte or Component Names | Nitroxoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H6 N2 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 8-Quinolinol, 5-nitro-, 5-(Hydroxyl[oxido]amino)-8-hydroxyquinoline, 5-Nitro-8-hydroxyquinoline, 5-Nitro-8-oxychinoline, 5-Nitro-8-oxyquinoline, 5-Nitro-8-quinolinol, 5-Nitrox, 5-Nitroxine, 5NOK, 8-Hydroxy-5-nitroquinoline, A 82, BAS 58, NSC 74947, NSC 74948, Nibiol, Nitroxolin, Nitroxoline, Noxibiol |
| InChI Formula | InChI=1S/C9H6N2O3/c12-8-4-3-7(11(13)14)6-2-1-5-10-9(6)8/h1-5,12H |
| SMILES | Oc1ccc(c2cccnc12)[N+](=O)[O-] |
| IUPAC Name | 5-nitroquinolin-8-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction