missing translation for 'onlineSavingsMsg'
Learn More

Nitroxoline, MedChemExpress

Product Code. 16412319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412319 Supplier Medchem Express Supplier No. HYB1159
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitroxoline is an antibiotic that has proven to be very effective at combating biofilm infections.

Nitroxoline is an antibiotic that has proven to be very effective at combating biofilm infections.

Specifications

Chemical Name or Material Nitroxoline
Physical Form Solid
CAS 4008-48-4
Color Light Green
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H6N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym 8-Hydroxy-5-nitroquinoline 5-Nitro-8-quinolinol
Solubility Information DMSO : 125 mg/mL (657.34 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=C([N+]([O-])=O)C=C1
Molecular Weight (g/mol) 190.16
Formula Weight 190.16
Percent Purity 99.51%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.