missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrotyrosine Antibody (EM-30), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-96130H
This item is not returnable.
View return policy
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Human, Mouse, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Nitrotyrosine | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| EM-30 | |
| Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Nitrotyrosine | |
| NO2-Tyr-CH2-Thyroglobulin | |
| 0.1 mL | |
| Primary | |
| Human, Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction