missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Nitrotough™ N230Y Series Medium Duty Oil Repellent Gloves

Product Code. 12861916
Change view
Click to view available options
:
Size: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12861916 Size: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12861916 Supplier Ansell Edmont™ Supplier No. C246100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal alternative to PVC and light leather gloves. Ansell Edmont™ Nitrotough™ N230Y Series Medium Duty Oil Repellent Gloves are good at abrasion resistance.

  • Mechanical protection gloves
  • Interlock 100% cotton liner
  • Elastic knit wrist for snug fit
  • Optimum comfort and flexibility
  • Excellent dexterity

Applications:

  • Assembly and finishing
  • General site and factory maintenance
  • Light metal fabrication and handling
  • Metal fabrication
  • Quality control
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Description Features OEKO -TEX™
Color Yellow
Exterior Finish 3/4 Dipped
Material Nitrile
Product Type Mechanical Protection Gloves
Size 10
Liner Material Cotton Jersey
Packaging Type 12 pairs in a bag, 12 bags in a carton
Series N230Y
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.