missing translation for 'onlineSavingsMsg'
Learn More

Nitrosyl Tetrafluoroborate, TRC

Product Code. 30475320
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30475320

Brand: Toronto Research Chemicals TRCN5500135G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrosyl Tetrafluoroborate
CAS 14635-75-7
Molecular Formula BF4NO
Quantity 5 g
Synonym Nitrosyl Borofluoride, Nitrosyl Fluoroborate, Nitrosonium Tetrafluoroborate, Nitrosonium Tetrafluoroborate, Nitroso Tetrafluoroborate, Nitrogen Fluoroborate Oxide, Nitrosyl Tetrafluoroborate, Nitrosyl Fluoborate
InChI Formula InChI=1S/BF4.NO/c2-1(3,4)5;1-2/q-1;+1
SMILES F[B-](F)(F)F.N#[O+]
IUPAC Name azanylidyneoxidanium;tetrafluoroborate
Molecular Weight (g/mol) 116.81
Formula Weight 117.0009
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.