missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrosyl Tetrafluoroborate, TRC
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Nitrosyl Tetrafluoroborate |
| CAS | 14635-75-7 |
| Molecular Formula | BF4NO |
| Quantity | 1 g |
| Synonym | Nitrosyl Borofluoride, Nitrosyl Fluoroborate, Nitrosonium Tetrafluoroborate, Nitrosonium Tetrafluoroborate, Nitroso Tetrafluoroborate, Nitrogen Fluoroborate Oxide, Nitrosyl Tetrafluoroborate, Nitrosyl Fluoborate |
| InChI Formula | InChI=1S/BF4.NO/c2-1(3,4)5;1-2/q-1;+1 |
| SMILES | F[B-](F)(F)F.N#[O+] |
| IUPAC Name | azanylidyneoxidanium;tetrafluoroborate |
| Molecular Weight (g/mol) | 116.81 |
| Formula Weight | 117.0009 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction