missing translation for 'onlineSavingsMsg'
Learn More

Nitrosamine Mixture 252 1000 μg/mL in Ethanol, Dr. Ehrenstorfer

Product Code. 30327612
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327612 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327612 Supplier Dr. Ehrenstorfer Supplier No. DREA50000252ET
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl-N-nitroso-2-propanamine∣N-Nitroso-diethylamine∣N-Nitrosodiisopropylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-butylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-N-methyl-4-aminobutyric Acid∣N-Nitroso-N-methylaniline
CAS 16339-04-1, 55-18-5, 601-77-4, 62-75-9, 924-16-3, 621-64-7
Analyte or Component Names N-Ethyl-N-nitroso-2-propanamine∣N-Nitroso-diethylamine∣N-Nitrosodiisopropylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-butylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-N-methyl-4-aminobutyric Acid∣N-Nitroso-N-methylaniline
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Ethanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) N-Ethyl-N-nitroso-2-propanamine 1000 μg/mL∣N-Nitroso-diethylamine 1000 μg/mL∣N-Nitrosodiisopropylamine 1000 μg/mL∣N-Nitrosodimethylamine 1000 μg/mL∣N-Nitroso-di-n-butylamine 1000 μg/mL∣N-Nitroso-di-n-propylamine 1000 μg/mL∣N-Nitroso-N-methyl-4-aminobutyric Acid 1000 μg/mL∣N-Nitroso-N-methylaniline 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.