missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrosamine Mixture 251 1000 μg/mL in Ethanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-nitroso-1-hexadecylamine∣N-Methyl-N-nitroso-1-octadecylamine∣N-Nitrosobis(2-hydroxypropyl)amine∣N-Nitrosodibenzylamine∣N-Nitrosodicyclohexylamine∣N-Nitrosodiisobutylamine∣N-Nitrosodiisononylamine∣N-Nitroso-diphenylamine∣N-Nitroso-N-ethylaniline∣N-Nitroso-N-methylbenzylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| CAS | 98648-34-1, 69112-94-3, 53609-64-6, 5336-53-8, 947-92-2, 997-95-5 |
| Analyte or Component Names | N-Methyl-N-nitroso-1-hexadecylamine∣N-Methyl-N-nitroso-1-octadecylamine∣N-Nitrosobis(2-hydroxypropyl)amine∣N-Nitrosodibenzylamine∣N-Nitrosodicyclohexylamine∣N-Nitrosodiisobutylamine∣N-Nitrosodiisononylamine∣N-Nitroso-diphenylamine∣N-Nitroso-N-ethylaniline∣N-Nitroso-N-methylbenzylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Ethanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | N-Methyl-N-nitroso-1-hexadecylamine 1000 μg/mL∣N-Methyl-N-nitroso-1-octadecylamine 1000 μg/mL∣N-Nitrosobis(2-hydroxypropyl)amine 1000 μg/mL∣N-Nitrosodibenzylamine 1000 μg/mL∣N-Nitrosodicyclohexylamine 1000 μg/mL∣N-Nitrosodiisobutylamine 1000 μg/mL∣N-Nitrosodiisononylamine 1000 μg/mL∣N-Nitroso-diphenylamine 1000 μg/mL∣N-Nitroso-N-ethylaniline 1000 μg/mL∣N-Nitroso-N-methylbenzylamine 1000 μg/mL∣N-Nitrosopiperidine 1000 μg/mL∣N-Nitrosopyrrolidine 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?