missing translation for 'onlineSavingsMsg'
Learn More

Nitroprusside disodium dihydrate, MedChemExpress

Product Code. 16468298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468298 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468298 Supplier Medchem Express Supplier No. HYA0119
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitroprusside disodium dehydrate (Sodium nitroprusside dihydrate) is a vasodilator that available for the research of acute hypertension, heart failure. Nitroprusside disodium dehydrate induces autophagy in glutathione-depleted osteoblasts. Nitroprusside disodium dehydrate acts as a nitric oxide (NO) donor in a rat intestinal ischemia reperfusion model.

Nitroprusside disodium dehydrate (Sodium nitroprusside dihydrate) is a vasodilator that available for the research of acute hypertension, heart failure. Nitroprusside disodium dehydrate induces autophagy in glutathione-depleted osteoblasts. Nitroprusside disodium dehydrate acts as a nitric oxide (NO) donor in a rat intestinal ischemia reperfusion model.

Specifications

Chemical Name or Material Nitroprusside disodium dihydrate
Physical Form Solid
CAS 13755-38-9
Color Red
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C5H4FeN6Na2O3
For Use With (Application) COVID-19-immunoregulation
Synonym Sodium nitroprusside dihydrate Sodium Nitroferricyanide(III) Dihydrate
Solubility Information H2O : 100 mg/mL (335.63 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (167.81 mM; Need ultrasonic)
SMILES [Na2[Fe(CN)5NO].2H2O]
Molecular Weight (g/mol) 297.95
Formula Weight 297.95
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.