missing translation for 'onlineSavingsMsg'
Learn More

Nitronium Tetrafluoroborate (Technical Grade), TRC

Product Code. 30480554
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30480554 100 mg -
30470839 50 mg 50mg
30473524 500 mg -
3 options
This item is not returnable. View return policy

Product Code. 30480554

Brand: Toronto Research Chemicals TRCN496868100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Formula Weight 132.9958
InChI Formula InChI=1S/BF4.NO2/c2-1(3,4)5;2-1-3/q-1;+1
IUPAC Name nitronium;tetrafluoroborate
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
SMILES F[B-](F)(F)F.O=[N+]=O
Synonym Nitryl Fluoborate, Nitryl tetrafluoroborate, Nitronium Fluoroborate, Nitronium Tetrafluoroborate, Nitronium Tetrafluoroborate, Nitryl Tetrafluoroborate, Nitryl Tetrafluoroborate
Chemical Name or Material Nitronium Tetrafluoroborate

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.